Assay Design
Gene Transcripts
- Gene Symbol
- kal-1
- Entrez Gene ID
-
173282
- Gene Name
- human KALlmann syndrome homolog
- Gene Aliases
-
CELE_K03D10.1
- Chromosome Location
-
Chr.I: 14128587 - 14142207 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061067.8 |
NP_493468.2 |
2 - 3 |
191 |
73 |
View Full Product Details