Assay Design



Gene Transcripts

Gene Symbol
exoc-8
Entrez Gene ID
173320
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.2
Chromosome Location
Chr.I: 14634449 - 14642574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026531.5 NP_001021702.1 4 - 5 908 111

View Full Product Details