Assay Design



Gene Transcripts

Gene Symbol
hmp-2
Entrez Gene ID
173338
Gene Name
protein-coding
Gene Aliases
CELE_K05C4.6
Chromosome Location
Chr.I: 14738882 - 14742671 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265496.3 NP_001252425.1 3 - 4 783 69
NM_001265497.3 NP_001252426.1 2 - 3 724 69
GenBank mRNA AF016853.1 - 2 - 3 749 69

View Full Product Details