Assay Design



Gene Transcripts

Gene Symbol
sur-2
Entrez Gene ID
173345
Gene Name
protein-coding
Gene Aliases
CELE_F39B2.4
Chromosome Location
Chr.I: 14777137 - 14787804 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026292.5 NP_001021463.1 4 - 5 964 108
NM_061174.6 NP_493575.2 4 - 5 965 108
GenBank mRNA U33051.1 - 4 - 5 966 108

View Full Product Details