Assay Design
Gene Transcripts
- Gene Symbol
- tre-5
- Entrez Gene ID
-
173388
- Gene Name
- Trehalase
- Gene Aliases
-
CELE_C23H3.7
- Chromosome Location
-
Chr.II: 52997 - 55304 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061248.5 |
NP_493649.2 |
1 - 2 |
153 |
67 |
GenBank mRNA |
AJ512339.1 |
- |
1 - 2 |
175 |
67 |
View Full Product Details