Assay Design



Gene Transcripts

Gene Symbol
pqn-85
Entrez Gene ID
173410
Gene Name
protein-coding
Gene Aliases
CELE_Y43H11AL.3
Chromosome Location
Chr.II: 240705 - 270669 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377684.3 NP_001364775.1 11 - 12 2303 183

View Full Product Details