Assay Design
Gene Transcripts
- Gene Symbol
- col-69
- Entrez Gene ID
-
186819
- Gene Name
- Col_cuticle_N domain-containing protein
- Gene Aliases
-
CELE_K01A2.7
- Chromosome Location
-
Chr.II: 303881 - 305745 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061301.3 |
NP_493702.1 |
1 - 2 |
385 |
77 |
View Full Product Details