Assay Design



Gene Transcripts

Gene Symbol
nhl-3
Entrez Gene ID
173452
Gene Name
protein-coding
Gene Aliases
CELE_W04H10.3
Chromosome Location
Chr.II: 595187 - 602120 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047352.4 NP_001040817.2 7 - 8 1007 63
NM_001047353.6 NP_001040818.2 4 - 5 707 63
NM_001316864.3 NP_001303793.1 1 - 2 248 63

View Full Product Details