Assay Design
Gene Transcripts
- Gene Symbol
- thoc-1
- Entrez Gene ID
-
173460
- Gene Name
- THO Complex (transcription factor/nuclear export) subunit
- Gene Aliases
-
CELE_T02H6.2
- Chromosome Location
-
Chr.II: 702107 - 708608 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061395.3 |
NP_493796.2 |
14 - 15 |
1690 |
81 |
View Full Product Details