Assay Design
Gene Transcripts
- Gene Symbol
- fbxc-31
- Entrez Gene ID
-
173472
- Gene Name
- F-box C protein
- Gene Aliases
-
CELE_R07C3.9
- Chromosome Location
-
Chr.II: 911980 - 913052 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061447.4 |
NP_493848.1 |
1 - 2 |
101 |
104 |
View Full Product Details