Assay Design
Gene Transcripts
- Gene Symbol
- fbxc-33
- Entrez Gene ID
-
173473
- Gene Name
- F-box C protein
- Gene Aliases
-
CELE_R07C3.8
- Chromosome Location
-
Chr.II: 913672 - 915219 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061448.4 |
NP_493849.1 |
2 - 3 |
1034 |
66 |
View Full Product Details