Assay Design
Gene Transcripts
- Gene Symbol
- btb-10
- Entrez Gene ID
-
173489
- Gene Name
- BTB domain-containing protein
- Gene Aliases
-
CELE_K02E7.9
- Chromosome Location
-
Chr.II: 1059662 - 1060802 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061501.4 |
NP_493902.1 |
1 - 2 |
156 |
79 |
View Full Product Details