Assay Design



Gene Transcripts

Gene Symbol
snf-3
Entrez Gene ID
173493
Gene Name
Sodium-and chloride-dependent betaine transporter
Gene Aliases
CELE_T13B5.1
Chromosome Location
Chr.II: 1092006 - 1097905 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061509.9 NP_493910.2 7 - 8 1523 87
GenBank mRNA DQ118731.1 - 7 - 8 1565 87
DQ482733.1 - 7 - 8 1545 87

View Full Product Details