Assay Design
Gene Transcripts
- Gene Symbol
- srw-66
- Entrez Gene ID
-
190359
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y57G7A.4
- Chromosome Location
-
Chr.II: 1269540 - 1271914 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061535.3 |
NP_493936.2 |
4 - 5 |
817 |
116 |
View Full Product Details