Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_061565.6 | NP_493966.1 | 5 - 6 | 1286 | 76 |
NM_061566.7 | NP_493967.1 | 4 - 5 | 951 | 76 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001381330.2 | NP_001368658.1 | 7 - 8 | 1834 | 76 |