Assay Design
Gene Transcripts
- Gene Symbol
- math-26
- Entrez Gene ID
-
173546
- Gene Name
- MATH (meprin-associated Traf homology) domain containing
- Gene Aliases
-
CELE_F36H5.1
- Chromosome Location
-
Chr.II: 1771647 - 1774955 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061674.6 |
NP_494075.1 |
10 - 11 |
2057 |
136 |
View Full Product Details