Assay Design



Gene Transcripts

Gene Symbol
math-13
Entrez Gene ID
182657
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_C16C4.3
Chromosome Location
Chr.II: 1875958 - 1877237 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377702.1 NP_001364700.1 1 - 2 101 100

View Full Product Details