Assay Design
Gene Transcripts
- Gene Symbol
- sdz-28
- Entrez Gene ID
-
173569
- Gene Name
- SKN-1 Dependent Zygotic transcript
- Gene Aliases
-
CELE_R52.1
- Chromosome Location
-
Chr.II: 2131070 - 2132475 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061791.4 |
NP_494192.1 |
4 - 5 |
802 |
152 |
View Full Product Details