Assay Design
Gene Transcripts
- Gene Symbol
- cct-3
- Entrez Gene ID
-
173581
- Gene Name
- T-complex protein 1 subunit gamma
- Gene Aliases
-
CELE_F54A3.3
- Chromosome Location
-
Chr.II: 2233540 - 2243539 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061817.6 |
NP_494218.2 |
4 - 5 |
776 |
85 |
View Full Product Details