Assay Design
Gene Transcripts
- Gene Symbol
- mnat-1
- Entrez Gene ID
-
173602
- Gene Name
- CDK-activating kinase assembly factor MAT1
- Gene Aliases
-
CELE_F53G2.7
- Chromosome Location
-
Chr.II: 2473512 - 2479091 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383802.1 |
|
8 - 9 |
1164 |
72 |
- Gene Symbol
- F53G2.12
- Entrez Gene ID
-
36805016
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F53G2.12
- Chromosome Location
-
Chr.II: 2473520 - 2479090 on Build WBcel235
View Full Product Details