Assay Design
Gene Transcripts
- Gene Symbol
- sri-37
- Entrez Gene ID
-
191911
- Gene Name
- Serpentine Receptor, class I
- Gene Aliases
-
CELE_D2062.2
- Chromosome Location
-
Chr.II: 2629570 - 2631231 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061910.2 |
NP_494311.2 |
3 - 4 |
494 |
87 |
View Full Product Details