Assay Design



Gene Transcripts

Gene Symbol
set-11
Entrez Gene ID
185242
Gene Name
SET (trithorax/polycomb) domain containing
Gene Aliases
CELE_F34D6.4
Chromosome Location
Chr.II: 2690472 - 2692576 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377732.1 NP_001364763.1 4 - 5 404 71

View Full Product Details