Assay Design
Gene Transcripts
- Gene Symbol
- fut-4
- Entrez Gene ID
-
187342
- Gene Name
- Glyco_tran_10_N domain-containing protein
- Gene Aliases
-
CELE_K12H6.3
- Chromosome Location
-
Chr.II: 2819435 - 2822535 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061971.5 |
NP_494372.2 |
5 - 6 |
569 |
85 |
GenBank mRNA |
AJ505020.1 |
- |
5 - 6 |
510 |
85 |
View Full Product Details