Assay Design
Gene Transcripts
- Gene Symbol
- sri-28
- Entrez Gene ID
-
191903
- Gene Name
- Serpentine Receptor, class I
- Gene Aliases
-
CELE_B0454.3
- Chromosome Location
-
Chr.II: 3048678 - 3051153 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062032.5 |
NP_494433.1 |
1 - 2 |
161 |
123 |
View Full Product Details