Assay Design
Gene Transcripts
- Gene Symbol
- pqn-48
- Entrez Gene ID
-
173743
- Gene Name
- Prion-like-(Q/N-rich)-domain-bearing protein
- Gene Aliases
-
CELE_K07D4.8
- Chromosome Location
-
Chr.II: 4038868 - 4043070 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062310.6 |
NP_494711.1 |
2 - 3 |
339 |
181 |
View Full Product Details