Assay Design



Gene Transcripts

Gene Symbol
rhgf-2
Entrez Gene ID
173748
Gene Name
protein-coding
Gene Aliases
CELE_T08H4.1
Chromosome Location
Chr.II: 4100785 - 4127849 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381382.1 NP_001368191.1 12 - 13 3273 88
NM_001381384.1 NP_001367992.1 12 - 13 3004 88
NM_001383815.1 12 - 13 3007 88
GenBank mRNA JQ763434.1 - 11 - 12 2847 88

View Full Product Details