Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_074154.5 | NP_506555.2 | 3 - 4 | 669 | 106 |
NM_182385.4 | NP_872185.2 | 3 - 4 | 575 | 106 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001028771.4 | NP_001023942.1 | 8 - 9 | 2016 | 106 |