Assay Design
Gene Transcripts
- Gene Symbol
- nlg-1
- Entrez Gene ID
-
181484
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C40C9.5
- Chromosome Location
-
Chr.X: 13625373 - 13631172 on Build WBcel235
- Gene Symbol
- Y12A6A.1
- Entrez Gene ID
-
181482
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y12A6A.1
- Chromosome Location
-
Chr.X: 13619938 - 13631150 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383667.1 |
|
12 - 13 |
2189 |
83 |
View Full Product Details