Assay Design



Gene Transcripts

Gene Symbol
nlg-1
Entrez Gene ID
181484
Gene Name
protein-coding
Gene Aliases
CELE_C40C9.5
Chromosome Location
Chr.X: 13625373 - 13631172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270296.3 NP_001257225.1 1 - 2 119 76
NM_001270297.3 NP_001257226.1 1 - 2 42 76
NM_001313376.3 NP_001300305.1 1 - 2 42 76
NM_001313377.3 NP_001300306.1 1 - 2 42 76
NM_001373559.4 NP_001359729.1 1 - 2 42 76
NM_077882.5 NP_510283.1 1 - 2 113 76
NM_182454.4 NP_872254.1 1 - 2 113 76

View Full Product Details