Assay Design



Gene Transcripts

Gene Symbol
gpa-11
Entrez Gene ID
173759
Gene Name
protein-coding
Gene Aliases
CELE_C16A11.1
Chromosome Location
Chr.II: 4247197 - 4255577 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267159.1 NP_001254088.1 5 - 6 951 135
NM_001267160.2 NP_001254089.1 5 - 6 940 135
GenBank mRNA AY008133.1 - 1 - 2 453 135

View Full Product Details