Assay Design



Gene Transcripts

Gene Symbol
twk-2
Entrez Gene ID
188452
Gene Name
protein-coding
Gene Aliases
CELE_T12C9.3
Chromosome Location
Chr.II: 4441103 - 4453151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001364060.1 NP_001350986.1 9 - 10 1100 101
NM_001364061.1 NP_001350985.1 9 - 10 1100 101
NM_001364062.1 NP_001350987.1 9 - 10 1100 101
NM_001364063.2 NP_001350984.1 9 - 10 1100 101
NM_001364064.3 NP_001350988.1 9 - 10 1100 101
NM_062385.7 NP_494786.5 9 - 10 1100 101

View Full Product Details