Assay Design
Gene Transcripts
- Gene Symbol
- sre-40
- Entrez Gene ID
-
188086
- Gene Name
- Serpentine Receptor, class E (epsilon)
- Gene Aliases
-
CELE_T05A7.8
- Chromosome Location
-
Chr.II: 4681715 - 4683793 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062429.3 |
NP_494830.2 |
2 - 3 |
538 |
123 |
View Full Product Details