Assay Design



Gene Transcripts

Gene Symbol
rig-6
Entrez Gene ID
173828
Gene Name
protein-coding
Gene Aliases
CELE_C33F10.5
Chromosome Location
Chr.II: 4764147 - 4812086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026843.7 NP_001022014.1 5 - 6 938 135
NR_151554.1 2 - 3 150 135

View Full Product Details