Assay Design
Gene Transcripts
- Gene Symbol
- gst-8
- Entrez Gene ID
-
184364
- Gene Name
- Probable glutathione S-transferase 8
- Gene Aliases
-
CELE_F11G11.1
- Chromosome Location
-
Chr.II: 4883712 - 4884602 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062483.3 |
NP_494884.1 |
1 - 2 |
175 |
88 |
View Full Product Details