Assay Design



Gene Transcripts

Gene Symbol
ulp-2
Entrez Gene ID
173859
Gene Name
protein-coding
Gene Aliases
CELE_Y38A8.3
Chromosome Location
Chr.II: 4955450 - 4959876 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318311.3 NP_001305240.1 5 - 5 610 69
NM_062513.5 NP_494914.1 4 - 5 610 69

View Full Product Details