Assay Design
Gene Transcripts
- Gene Symbol
- ulp-2
- Entrez Gene ID
-
173859
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y38A8.3
- Chromosome Location
-
Chr.II: 4955450 - 4959876 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062513.5 |
NP_494914.1 |
6 - 7 |
923 |
103 |
View Full Product Details