Assay Design
Gene Transcripts
- Gene Symbol
- ptr-6
- Entrez Gene ID
-
173898
- Gene Name
- SSD domain-containing protein
- Gene Aliases
-
CELE_C54A12.1
- Chromosome Location
-
Chr.II: 5242442 - 5246837 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062585.6 |
NP_494986.2 |
6 - 7 |
1140 |
63 |
View Full Product Details