Assay Design



Gene Transcripts

Gene Symbol
gcy-12
Entrez Gene ID
173902
Gene Name
Receptor-type guanylate cyclase gcy-12
Gene Aliases
CELE_F08B1.2
Chromosome Location
Chr.II: 5312046 - 5319617 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062594.3 NP_494995.2 13 - 14 4585 68
NR_134556.1 13 - 14 4843 68

View Full Product Details