Assay Design
Gene Transcripts
- Gene Symbol
- pink-1
- Entrez Gene ID
-
173918
- Gene Name
- Serine/threonine-protein kinase pink-1, mitochondrial
- Gene Aliases
-
CELE_EEED8.9
- Chromosome Location
-
Chr.II: 5393292 - 5395694 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062616.6 |
NP_495017.1 |
4 - 5 |
595 |
87 |
View Full Product Details