Assay Design
Gene Transcripts
- Gene Symbol
- mog-5
- Entrez Gene ID
-
173920
- Gene Name
- Probable pre-mRNA-splicing factor ATP-dependent RNA helicase mog-5
- Gene Aliases
-
CELE_EEED8.5
- Chromosome Location
-
Chr.II: 5399538 - 5405810 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062618.7 |
NP_495019.1 |
1 - 2 |
166 |
92 |
GenBank mRNA |
AF286899.1 |
- |
1 - 2 |
173 |
92 |
View Full Product Details