Assay Design



Gene Transcripts

Gene Symbol
acdh-2
Entrez Gene ID
173940
Gene Name
protein-coding
Gene Aliases
CELE_C17C3.12
Chromosome Location
Chr.II: 5544778 - 5547492 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356838.3 NP_001343623.1 3 - 4 682 82
NM_001356841.4 NP_001343742.1 - 632 82
NM_001373716.3 NP_001360428.1 3 - 4 679 82
NM_001383838.1 3 - 4 499 82
NM_062665.8 NP_495066.2 3 - 4 682 82

View Full Product Details