Assay Design
Gene Transcripts
- Gene Symbol
- zyg-1
- Entrez Gene ID
-
173956
- Gene Name
- Probable serine/threonine-protein kinase zyg-1
- Gene Aliases
-
CELE_F59E12.2
- Chromosome Location
-
Chr.II: 5649748 - 5654422 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062702.6 |
NP_495103.1 |
2 - 3 |
151 |
87 |
GenBank mRNA |
AF285179.1 |
- |
2 - 3 |
173 |
87 |
View Full Product Details