Assay Design



Gene Transcripts

Gene Symbol
ifb-1
Entrez Gene ID
173976
Gene Name
protein-coding
Gene Aliases
CELE_F10C1.2
Chromosome Location
Chr.II: 5765612 - 5772126 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383849.1 3 - 4 541 58
NM_062736.4 NP_495137.1 3 - 4 434 58

View Full Product Details