Assay Design
Gene Transcripts
- Gene Symbol
- ifb-1
- Entrez Gene ID
-
173976
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F10C1.2
- Chromosome Location
-
Chr.II: 5765612 - 5772126 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062736.4 |
NP_495137.1 |
1 - 2 |
61 |
59 |
View Full Product Details