Assay Design
Gene Transcripts
- Gene Symbol
- ins-3
- Entrez Gene ID
-
191684
- Gene Name
- Probable insulin-like peptide beta-type 3
- Gene Aliases
-
CELE_ZK75.3
- Chromosome Location
-
Chr.II: 5990099 - 5990770 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062794.8 |
NP_495195.1 |
1 - 2 |
297 |
117 |
View Full Product Details