Assay Design
Gene Transcripts
- Gene Symbol
- ran-5
- Entrez Gene ID
-
174011
- Gene Name
- RanBD1 domain-containing protein
- Gene Aliases
-
CELE_R12C12.2
- Chromosome Location
-
Chr.II: 6048103 - 6052538 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062807.8 |
NP_495208.1 |
1 - 2 |
76 |
92 |
View Full Product Details