Assay Design



Gene Transcripts

Gene Symbol
bpl-1
Entrez Gene ID
174039
Gene Name
protein-coding
Gene Aliases
CELE_F13H8.10
Chromosome Location
Chr.II: 6274115 - 6279720 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383861.1 8 - 9 1397 95
NM_062856.7 NP_495257.3 8 - 9 1441 95

View Full Product Details