Assay Design
Gene Transcripts
- Gene Symbol
- nhr-22
- Entrez Gene ID
-
174066
- Gene Name
- Nuclear hormone receptor family member nhr-22
- Gene Aliases
-
CELE_K06A1.4
- Chromosome Location
-
Chr.II: 6446195 - 6449815 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062897.6 |
NP_495298.1 |
1 - 2 |
366 |
106 |
View Full Product Details