Assay Design



Gene Transcripts

Gene Symbol
snr-3
Entrez Gene ID
174072
Gene Name
protein-coding
Gene Aliases
CELE_T28D9.10
Chromosome Location
Chr.II: 6488829 - 6489761 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330951.3 NP_001317766.1 1 - 2 98 67
NM_062905.7 NP_495306.1 2 - 3 193 67

View Full Product Details