Assay Design
Gene Transcripts
- Gene Symbol
- abch-1
- Entrez Gene ID
-
174078
- Gene Name
- ABC transporter domain-containing protein
- Gene Aliases
-
CELE_C56E6.5
- Chromosome Location
-
Chr.II: 6528929 - 6531627 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062919.5 |
NP_495320.1 |
2 - 3 |
326 |
71 |
View Full Product Details